Tisk…

4-(Difluoromethyl)tetrahydro-2H-pyran

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Difluoromethyl)oxane

PC52073-1G PC52073-250MG PC52073-5G
APOSPC52073-1GEA 3640 CZK
APOSPC52073-1G APOSPC52073-250MG APOSPC52073-5G
4-(Difluoromethyl)tetrahydro-2H-pyran
4-(Difluoromethyl)tetrahydro-2H-pyran
Vzorec: C₆H₁₀F₂O
MW: 136,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28014019
CAS číslo: 1820618-46-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR