Tisk…

1-Fluoro-4-(pentafluoroethyl)benzene 97%

Dodavatel: Apollo Scientific

PC520721-10G PC520721-1G PC520721-5G
APOSPC520721-10GEA 43310 CZK
APOSPC520721-10G APOSPC520721-1G APOSPC520721-5G
1-Fluoro-4-(pentafluoroethyl)benzene 97%
1-Fluoro-4-(pentafluoroethyl)benzene
Vzorec: C₈H₄F₆
MW: 214,108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16140153
CAS číslo: 2396-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR