Tisk…

4-Fluoro-2-methoxybenzenesulphonyl chloride 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Fluoro-2-methoxybenzene-1-sulphonyl chloride, 2-(Chlorosulphonyl)-5-fluoroanisole

PC52070-1G PC52070-250MG
APOSPC52070-1GEA 25630 CZK
APOSPC52070-1G APOSPC52070-250MG
4-Fluoro-2-methoxybenzenesulphonyl chloride 95%
4-Fluoro-2-methoxybenzenesulphonyl chloride
Vzorec: C₇H₆ClFO₃S
MW: 224,637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185596
CAS číslo: 1214377-19-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR