2-(Trifluoromethyl)cyclopropanamine hydrochloride 95%
Dodavatel: Apollo Scientific
PC520694-1G
PC520694-250MG
PC520694-500MG
APOSPC520694-1GEA
30820
CZK
APOSPC520694-1G
APOSPC520694-250MG
APOSPC520694-500MG
2-(Trifluoromethyl)cyclopropanamine hydrochloride 95%
2-(Trifluoromethyl)cyclopropanamine hydrochloride
Vzorec:
C₄H₆F₃N·ClH MW: 161,553 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13806389 CAS číslo: 1172072-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...