Tisk…

2-Methyl-5-(trifluoromethyl)pyrazine 96%

Dodavatel: Apollo Scientific

PC520682-10G PC520682-5G
APOSPC520682-10GEA 23780 CZK
APOSPC520682-10G APOSPC520682-5G
2-Methyl-5-(trifluoromethyl)pyrazine 96%
2-Methyl-5-(trifluoromethyl)pyrazine
Vzorec: C₆H₅F₃N₂
MW: 162,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546943
CAS číslo: 1186195-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR