Tisk…

4,4-Difluoro-1-methylpiperidine 97%

Dodavatel: Apollo Scientific

PC520679-1G PC520679-5G
APOSPC520679-1GEA 3510 CZK
APOSPC520679-1G APOSPC520679-5G
4,4-Difluoro-1-methylpiperidine 97%
4,4-Difluoro-1-methylpiperidine
Vzorec: C₆H₁₁F₂N
Storage Temperature: Ambient
MDL Number: MFCD12546945
CAS číslo: 1186194-60-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR