Tisk…

3-(Trifluoromethoxy)phenylmethanesulphonyl chloride 97%

Dodavatel: Apollo Scientific

PC520678-1G PC520678-5G
APOSPC520678-1GEA 22880 CZK
APOSPC520678-1G APOSPC520678-5G
3-(Trifluoromethoxy)phenylmethanesulphonyl chloride 97%
3-(Trifluoromethoxy)phenylmethanesulphonyl chloride
Vzorec: C₈H₆ClF₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD11976109
CAS číslo: 1263274-68-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR