Ethyl 4-amino-2,6-difluorobenzoate 97%
Dodavatel: Apollo Scientific
PC520677-1G
PC520677-5G
APOSPC520677-1GEA
8630
CZK
APOSPC520677-1G
APOSPC520677-5G
Ethyl 4-amino-2,6-difluorobenzoate 97%
Ethyl 4-amino-2,6-difluorobenzoate
Vzorec:
C₉H₉F₂NO₂ MW: 201.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11973735 CAS číslo: 191469-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...