3-Fluoro-2-methoxybenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Chlorosulphonyl)-6-fluoroanisole
PC52066-1G
PC52066-250MG
APOSPC52066-1GEA
16530
CZK
APOSPC52066-1G
APOSPC52066-250MG
3-Fluoro-2-methoxybenzenesulphonyl chloride
3-Fluoro-2-methoxybenzenesulphonyl chloride
Vzorec:
C₇H₆ClFO₃S MW: 224,637 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185594 CAS číslo: 1049729-85-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...