Tisk…
2-Hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylic acid methyl ester hydrate 97%
Dodavatel: Apollo Scientific
PC520631-25G PC520631-5G
APOSPC520631-25GEA 28170 CZK
APOSPC520631-25G APOSPC520631-5G
2-Hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylic acid methyl ester hydrate 97%
2-Hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylic acid methyl ester hydrate
Vzorec: C₇H₅F₃N₂O₃·H₂O
MW: 240.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100556
CAS číslo: 886212-78-8