1-Chloro-2-fluoro-4-(2,2,2-trifluoroethyl)benzene 95%
Dodavatel: Apollo Scientific
PC520624-10G
PC520624-5G
APOSPC520624-10GEA
32800
CZK
APOSPC520624-10G
APOSPC520624-5G
1-Chloro-2-fluoro-4-(2,2,2-trifluoroethyl)benzene 95%
1-Chloro-2-fluoro-4-(2,2,2-trifluoroethyl)benzene
Vzorec:
C₈H₅ClF₄ MW: 212,572 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226696 CAS číslo: 1186194-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...