Tisk…

6-Bromo-2,4-dichloro-3-fluoroaniline 97%

Dodavatel: Apollo Scientific

PC520607-5G PC520607-250MG PC520607-1G
APOSPC520607-5GEA 22320 CZK
APOSPC520607-5G APOSPC520607-250MG APOSPC520607-1G
6-Bromo-2,4-dichloro-3-fluoroaniline 97%
6-Bromo-2,4-dichloro-3-fluoroaniline
Vzorec: C₆H₃BrCl₂FN
Storage Temperature: Ambient
MDL Number: MFCD11520111
CAS číslo: 1360438-57-8
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR