Tisk…

3-Fluoro-2-methoxy-5-(trifluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific

PC520597-1G PC520597-5G
APOSPC520597-1GEA 3510 CZK
APOSPC520597-1G APOSPC520597-5G
3-Fluoro-2-methoxy-5-(trifluoromethyl)pyridine 97%
3-Fluoro-2-methoxy-5-(trifluoromethyl)pyridine
Vzorec: C₇H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD10699120
CAS číslo: 1138011-20-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR