4-Cyclopropyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 95%
Dodavatel: Apollo Scientific
PC520573-1G
PC520573-5G
APOSPC520573-1GEA
9290
CZK
APOSPC520573-1G
APOSPC520573-5G
4-Cyclopropyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 95%
4-Cyclopropyl-2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Vzorec:
C₉H₇F₃N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08689831 CAS číslo: 914201-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...