(6-Fluoro-2-pyridyl)methanol 97%
Dodavatel: Apollo Scientific
Synonyms:
6-Fluoro-2-pyridinemethanol
PC520559-5G
APOSPC520559-5GEA
4900
CZK
APOSPC520559-5G
(6-Fluoro-2-pyridyl)methanol 97%
(6-Fluoro-2-pyridyl)methanol
Vzorec:
C₆H₆FNO MW: 127,116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07783827 CAS číslo: 315180-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...