Tisk…

5-Chloro-2-fluoro-1-(difluromethyl)benzene 95%

Dodavatel: Apollo Scientific

PC520557-10G PC520557-1G PC520557-5G
APOSPC520557-10GEA 30490 CZK
APOSPC520557-10G APOSPC520557-1G APOSPC520557-5G
5-Chloro-2-fluoro-1-(difluromethyl)benzene 95%
5-Chloro-2-fluoro-1-(difluromethyl)benzene
Vzorec: C₇H₄ClF₃
Storage Temperature: Ambient
MDL Number: MFCD07779540
CAS číslo: 63878-72-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR