Tisk…

3-Bromo-5-fluoro-4-methylaniline 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Amino-2-bromo-6-fluorotoluene, 3-Bromo-5-fluoro-p-toluidine

PC52052-1G PC52052-250MG PC52052-5G
APOSPC52052-1GEA 6390 CZK
APOSPC52052-1G APOSPC52052-250MG APOSPC52052-5G
3-Bromo-5-fluoro-4-methylaniline 98%
3-Bromo-5-fluoro-4-methylaniline
Vzorec: C₇H₇BrFN
MW: 204,04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025433
CAS číslo: 207110-35-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR