N-(4-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide 95%
Dodavatel: Apollo Scientific
PC520510-1G
PC520510-5G
APOSPC520510-1GEA
7970
CZK
APOSPC520510-1G
APOSPC520510-5G
N-(4-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide 95%
N-(4-Chlorophenyl)-1,1,1-trifluoro-methanesulphonamide
Vzorec:
C₇H₅ClF₃NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD00856978 CAS číslo: 23384-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...