Tisk…

3-Acetyl-3-fluoro-tetrahydrofuran-2-one 95%

Dodavatel: Apollo Scientific

PC520505-10G PC520505-1G PC520505-5G
APOSPC520505-10GEA 22550 CZK
APOSPC520505-10G APOSPC520505-1G APOSPC520505-5G
3-Acetyl-3-fluoro-tetrahydrofuran-2-one 95%
3-Acetyl-3-fluoro-tetrahydrofuran-2-one
Vzorec: C₆H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD00215829
CAS číslo: 63141-03-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR