1-Bromo-2-(difluoromethyl)-3-fluoro-4-methoxybenzene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC520328-1G
PC520328-250MG
APOSPC520328-1GEA
34060
CZK
APOSPC520328-1G
APOSPC520328-250MG
1-Bromo-2-(difluoromethyl)-3-fluoro-4-methoxybenzene 95%
1-Bromo-2-(difluoromethyl)-3-fluoro-4-methoxybenzene
Vzorec:
C₈H₆BrF₃O MW: 255.032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29767785 CAS číslo: 2092668-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...