2-(2,2,2-Trifluoroethoxy)nitrobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC5196-1G
PC5196-25G
PC5196-5G
APOSPC5196-1GEA
744
CZK
APOSPC5196-1G
APOSPC5196-25G
APOSPC5196-5G
2-(2,2,2-Trifluoroethoxy)nitrobenzene 97%
2-(2,2,2-Trifluoroethoxy)nitrobenzene
Vzorec:
C₈H₆F₃NO₃ MW: 221,133 g/mol Bod varu: 114 °C (3 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00077607 CAS číslo: 87014-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...