Tisk…

HFPO polymer allyl ether

Dodavatel: Apollo Scientific

PC51958-25G PC51958-5G
APOSPC51958-25GEA 17300 CZK
APOSPC51958-25G APOSPC51958-5G
HFPO polymer allyl ether
HFPO polymer allyl ether

(3000 - 4000 amu)

Hustota: 1.8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD09753871
CAS číslo: 631897-32-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR