Tisk…

2-(3-Fluoro-2-nitrophenyl)acetonitrile

Dodavatel: Apollo Scientific

PC51905-1G PC51905-5G
APOSPC51905-1GEA 2200 CZK
APOSPC51905-1G APOSPC51905-5G
2-(3-Fluoro-2-nitrophenyl)acetonitrile
2-(3-Fluoro-2-nitrophenyl)acetonitrile
Vzorec: C₈H₅FN₂O₂
MW: 180,136 g/mol
Bod tání: 72…73 °C
Storage Temperature: Ambient
MDL Number: MFCD13194393
CAS číslo: 105003-88-1
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR