6-Nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC51862-1G
PC51862-250MG
APOSPC51862-1GEA
2690
CZK
APOSPC51862-1G
APOSPC51862-250MG
6-Nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
6-Nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Vzorec:
C₉H₄F₃N₃O₄ MW: 275,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22018788 CAS číslo: 1483017-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...