2,3-Difluoro-4-(trifluoromethoxy)aniline 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC51845-1G
PC51845-250MG
PC51845-5G
APOSPC51845-1GEA
17360
CZK
APOSPC51845-1G
APOSPC51845-250MG
APOSPC51845-5G
2,3-Difluoro-4-(trifluoromethoxy)aniline 95%
2,3-Difluoro-4-(trifluoromethoxy)aniline
Vzorec:
C₇H₄F₅NO MW: 213.105 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371363 CAS číslo: 107570-54-7 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...