2-Bromo-6-(difluoromethyl)benzaldehyde 95%
Dodavatel: Apollo Scientific
PC51629-1G
PC51629-250MG
APOSPC51629-1GEA
18620
CZK
APOSPC51629-1G
APOSPC51629-250MG
2-Bromo-6-(difluoromethyl)benzaldehyde 95%
2-Bromo-6-(difluoromethyl)benzaldehyde
Vzorec:
C₈H₅BrF₂O MW: 235,026 g/mol Storage Temperature: Ambient |
CAS číslo:
1780690-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...