8-(Trifluoromethoxy)-4-hydroxyquinoline
Dodavatel: Apollo Scientific
Synonyms:
8-(Trifluoromethoxy)quinolin-4-ol
PC51523-1G
PC51523-250MG
APOSPC51523-1GEA
7410
CZK
APOSPC51523-1G
APOSPC51523-250MG
8-(Trifluoromethoxy)-4-hydroxyquinoline
8-(Trifluoromethoxy)-4-hydroxyquinoline
Vzorec:
C₁₀H₆F₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03407893 CAS číslo: 40516-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...