Tisk…

Methyl 6-bromo-2,3,4-trifluorobenzoate

Dodavatel: Apollo Scientific

PC51486-1G PC51486-250MG PC51486-5G
APOSPC51486-1GEA 2400 CZK
APOSPC51486-1G APOSPC51486-250MG APOSPC51486-5G
Methyl 6-bromo-2,3,4-trifluorobenzoate
Methyl 6-bromo-2,3,4-trifluorobenzoate
Vzorec: C₈H₄BrF₃O₂
MW: 269,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32647609
CAS číslo: 1525649-77-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR