Tisk…

1,3-Difluoro-2-iodo-5-(trifluoromethoxy)benzene 95%

Dodavatel: Apollo Scientific
Varování

PC51476-1G PC51476-250MG PC51476-5G
APOSPC51476-1GEA 992 CZK
APOSPC51476-1G APOSPC51476-250MG APOSPC51476-5G
1,3-Difluoro-2-iodo-5-(trifluoromethoxy)benzene 95%
1,3-Difluoro-2-iodo-5-(trifluoromethoxy)benzene
Vzorec: C₇H₂F₅IO
Storage Temperature: Ambient
MDL Number: MFCD32219443
CAS číslo: 2384319-66-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR