2,3-Difluoro-4-(trifluoromethoxy)phenol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC51437-1G
PC51437-250MG
PC51437-5G
APOSPC51437-1GEA
17760
CZK
APOSPC51437-1G
APOSPC51437-250MG
APOSPC51437-5G
2,3-Difluoro-4-(trifluoromethoxy)phenol 95%
2,3-Difluoro-4-(trifluoromethoxy)phenol
Vzorec:
C₇H₃F₅O₂ MW: 214,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32066749 CAS číslo: 158178-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...