Tisk…

2-Fluoro-5-(3-oxo-3H-isobenzofuran-1-ylidenemethyl)-benzonitrile 97%

Dodavatel: Apollo Scientific

PC51400-1G PC51400-25G PC51400-5G
APOSPC51400-1GEA 2240 CZK
APOSPC51400-1G APOSPC51400-25G APOSPC51400-5G
2-Fluoro-5-(3-oxo-3H-isobenzofuran-1-ylidenemethyl)-benzonitrile 97%
2-Fluoro-5-(3-oxo-3H-isobenzofuran-1-ylidenemethyl)-benzonitrile
Vzorec: C₁₆H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD13181869
CAS číslo: 763114-25-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR