2,3-Difluoro-4-(trifluoromethoxy)benzeneboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
[2,3-difluoro-4-(trifluoromethoxy)phenyl]boronic acid
PC51396-1G
PC51396-250MG
PC51396-5G
APOSPC51396-1GEA
14880
CZK
APOSPC51396-1G
APOSPC51396-250MG
APOSPC51396-5G
2,3-Difluoro-4-(trifluoromethoxy)benzeneboronic acid 95%
2,3-Difluoro-4-(trifluoromethoxy)benzeneboronic acid
Vzorec:
C₇H₄BF₅O₃ MW: 241.908 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29919696 CAS číslo: 158178-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...