5-Bromo-2,3,4-trifluorobenzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(5-Bromo-2,3,4-trifluorophenyl)methanol
PC51349-1G
PC51349-250MG
PC51349-5G
APOSPC51349-1GEA
1660
CZK
APOSPC51349-1G
APOSPC51349-250MG
APOSPC51349-5G
5-Bromo-2,3,4-trifluorobenzyl alcohol 95%
5-Bromo-2,3,4-trifluorobenzyl alcohol
Vzorec:
C₇H₄BrF₃O MW: 241,005 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12756768 CAS číslo: 511511-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...