Tisk…

[4-(1,1,2,2-Tetrafluoroethoxy)phenyl]boronic acid, pinacol ester 95%

Dodavatel: Apollo Scientific

PC51347-1G PC51347-250MG
APOSPC51347-1GEA 16340 CZK
APOSPC51347-1G APOSPC51347-250MG
[4-(1,1,2,2-Tetrafluoroethoxy)phenyl]boronic acid, pinacol ester 95%
[4-(1,1,2,2-Tetrafluoroethoxy)phenyl]boronic acid, pinacol ester
Vzorec: C₁₄H₁₇BF₄O₃
MW: 320.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996389
CAS číslo: 503309-12-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR