Tisk…

5-Amino-2-fluorophenylacetic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Carboxymethyl)-4-fluoroaniline

PC5134-1G PC5134-5G PC5134-10G PC5134-25G
APOSPC5134-1GEA 622 CZK
APOSPC5134-1G APOSPC5134-5G APOSPC5134-10G APOSPC5134-25G
5-Amino-2-fluorophenylacetic acid 98%
5-Amino-2-fluorophenylacetic acid
Vzorec: C₈H₈FNO₂
MW: 169,153 g/mol
Bod tání: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD03094259
CAS číslo: 518057-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR