Tisk…

1-(4-Fluorophenyl)-1,2,4-triazole-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

PC51335-1G PC51335-250MG
APOSPC51335-1GEA 20470 CZK
APOSPC51335-1G APOSPC51335-250MG
1-(4-Fluorophenyl)-1,2,4-triazole-3-carboxylic acid
1-(4-Fluorophenyl)-1,2,4-triazole-3-carboxylic acid
Vzorec: C₉H₆FN₃O₂
MW: 207,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28606409
CAS číslo: 1552231-28-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR