Tisk…

Ethyl 1-(4-fluorophenyl)-1,2,4-triazole-3-carboxylate

Dodavatel: Apollo Scientific
Varování

PC51334-1G PC51334-250MG
APOSPC51334-1GEA 14880 CZK
APOSPC51334-1G APOSPC51334-250MG
Ethyl 1-(4-fluorophenyl)-1,2,4-triazole-3-carboxylate
Ethyl 1-(4-fluorophenyl)-1,2,4-triazole-3-carboxylate
Vzorec: C₁₁H₁₀FN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD29905001
CAS číslo: 1613022-95-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR