Tisk…

2-Amino-4-fluoro-1,3-benzothiazole-6-carbonitrile 97%

Dodavatel: Apollo Scientific
Varování

PC51333-1G PC51333-250MG
APOSPC51333-1GEA 18650 CZK
APOSPC51333-1G APOSPC51333-250MG
2-Amino-4-fluoro-1,3-benzothiazole-6-carbonitrile 97%
2-Amino-4-fluoro-1,3-benzothiazole-6-carbonitrile
Vzorec: C₈H₄FN₃S
MW: 193,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705710
CAS číslo: 1427448-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR