Tisk…

5-Bromo-4-fluoro-1,3-benzothiazol-2-amine 99%

Dodavatel: Apollo Scientific
Varování

PC51332-1G PC51332-250MG
APOSPC51332-1GEA 18650 CZK
APOSPC51332-1G APOSPC51332-250MG
5-Bromo-4-fluoro-1,3-benzothiazol-2-amine 99%
5-Bromo-4-fluoro-1,3-benzothiazol-2-amine
Vzorec: C₇H₄BrFN₂S
MW: 247,087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705700
CAS číslo: 1427383-09-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR