Tisk…

2,5-Dibromo-4-fluoro-1,3-benzothiazole

Dodavatel: Apollo Scientific
Varování

PC51330-1G PC51330-250MG
APOSPC51330-1GEA 7380 CZK
APOSPC51330-1G APOSPC51330-250MG
2,5-Dibromo-4-fluoro-1,3-benzothiazole
2,5-Dibromo-4-fluoro-1,3-benzothiazole
Vzorec: C₇H₂Br₂FNS
MW: 310.969 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705942
CAS číslo: 1427426-59-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR