2-Bromo-4-fluoro-1,3-benzothiazole-6-carbonitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC51329-100MG
PC51329-1G
PC51329-250MG
APOSPC51329-100MGEA
3410
CZK
APOSPC51329-100MG
APOSPC51329-1G
APOSPC51329-250MG
2-Bromo-4-fluoro-1,3-benzothiazole-6-carbonitrile 97%
2-Bromo-4-fluoro-1,3-benzothiazole-6-carbonitrile
Vzorec:
C₈H₂BrFN₂S MW: 257,082 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23705953 CAS číslo: 1427399-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...