2-(2,2-Dimethylpropanoylamino)-6-(trifluoromethyl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC51317-1G
PC51317-250MG
APOSPC51317-1GEA
7440
CZK
APOSPC51317-1G
APOSPC51317-250MG
2-(2,2-Dimethylpropanoylamino)-6-(trifluoromethyl)benzoic acid
2-(2,2-Dimethylpropanoylamino)-6-(trifluoromethyl)benzoic acid
Vzorec:
C₁₃H₁₄F₃NO₃ MW: 289.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00097847 CAS číslo: 256508-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...