5-Amino-2-fluorobenzamide 95%
Dodavatel: Apollo Scientific
PC5125-5G
PC5125-25G
APOSPC5125-5GEA
9590
CZK
APOSPC5125-5G
APOSPC5125-25G
5-Amino-2-fluorobenzamide 95%
5-Amino-2-fluorobenzamide
Vzorec:
C₇H₇FN₂O MW: 154,142 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094256 CAS číslo: 518057-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...