Tisk…

4-Bromo-5-fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-4-fluoro-2-nitrophenyl 2,2,2-trifluoroethyl ether, 1-Bromo-2-fluoro-4-nitro-5-(2,2,2-trifluoroethoxy)benzene

PC51229-1G PC51229-5G
APOSPC51229-1GEA 11640 CZK
APOSPC51229-1G APOSPC51229-5G
4-Bromo-5-fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene
4-Bromo-5-fluoro-2-(2,2,2-trifluoroethoxy)nitrobenzene
Vzorec: C₈H₄BrF₄NO₃
MW: 318,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28155020
CAS číslo: 1820716-82-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR