2,4-Difluoro-5-nitrobenzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC51227-1G
PC51227-250MG
PC51227-5G
APOSPC51227-1GEA
9330
CZK
APOSPC51227-1G
APOSPC51227-250MG
APOSPC51227-5G
2,4-Difluoro-5-nitrobenzaldehyde 95%
2,4-Difluoro-5-nitrobenzaldehyde
Vzorec:
C₇H₃F₂NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11857779 CAS číslo: 127228-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...