4-Chloro-5-(hydroxymethyl)-2-(trifluoromethyl)pyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
[4-Chloro-6-(trifluoromethyl)pyridin-3-yl]methanol, 4-Chloro-6-(trifluoromethyl)nicotinyl alcohol
PC51217-1G
PC51217-250MG
APOSPC51217-1GEA
33730
CZK
APOSPC51217-1G
APOSPC51217-250MG
4-Chloro-5-(hydroxymethyl)-2-(trifluoromethyl)pyridine
4-Chloro-5-(hydroxymethyl)-2-(trifluoromethyl)pyridine
Vzorec:
C₇H₅ClF₃NO MW: 211,569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24369748 CAS číslo: 1431842-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...