5-Amino-4-hydroxy-2-(trifluoromethyl)pyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-2-(trifluoromethyl)pyridin-4-ol, 4-Hydroxy-6-(trifluoromethyl)pyridin-3-amine
PC51215-100MG
PC51215-50MG
APOSPC51215-100MGEA
8900
CZK
APOSPC51215-100MG
APOSPC51215-50MG
5-Amino-4-hydroxy-2-(trifluoromethyl)pyridine
5-Amino-4-hydroxy-2-(trifluoromethyl)pyridine
Vzorec:
C₆H₅F₃N₂O MW: 178,112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189822 CAS číslo: 1196153-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...