2-(Difluoromethyl)isonicotinaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Difluoromethyl)-4-formylpyridine, 2-(Difluoromethyl)pyridine-4-carboxaldehyde
PC51203-1G
PC51203-250MG
PC51203-100MG
APOSPC51203-1GEA
34060
CZK
APOSPC51203-1G
APOSPC51203-250MG
APOSPC51203-100MG
2-(Difluoromethyl)isonicotinaldehyde
2-(Difluoromethyl)isonicotinaldehyde
Vzorec:
C₇H₅F₂NO MW: 157,118 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18256101 CAS číslo: 1211540-67-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...