Tisk…

N-[(1R)-1-(3-Fluorophenyl)ethyl]cyclopropanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC512028-1G PC512028-250MG
APOSPC512028-1GEA 12200 CZK
APOSPC512028-1G APOSPC512028-250MG
N-[(1R)-1-(3-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
N-[(1R)-1-(3-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
Vzorec: C₁₁H₁₄FN·ClH
MW: 215,695 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32066862
CAS číslo: 2414145-16-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR