Tisk…

2-(Difluoromethyl)isonicotinonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Difluoromethyl)pyridine-4-carbonitrile, 4-Cyano-2-(difluoromethyl)pyridine

PC51202-1G PC51202-250MG
APOSPC51202-1GEA 25630 CZK
APOSPC51202-1G APOSPC51202-250MG
2-(Difluoromethyl)isonicotinonitrile
2-(Difluoromethyl)isonicotinonitrile
Vzorec: C₇H₄F₂N₂
MW: 154,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256051
CAS číslo: 1211528-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR